2-(Benzyloxy)-1-bromo-4-fluorobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Benzyloxy-1-bromo-4-fluorobenzene

071188-5G
102519-586EA 950.77 USD
102519-586
2-(Benzyloxy)-1-bromo-4-fluorobenzene ≥97%
2-(Benzyloxy)-1-bromo-4-fluorobenzene

Hazard: Irritant

Formula: C₁₃H₁₀BrFO
MW: 281.13 g/mol
MDL Number: MFCD09033866
CAS Number: 202857-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR