4-Bromo-5-methyl-2-nitroaniline ≥97%

Supplier: Matrix Scientific

071173-5G
102519-562EA 952.31 USD
102519-562
4-Bromo-5-methyl-2-nitroaniline ≥97%
4-Bromo-5-methyl-2-nitroaniline

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
Boiling Pt: 335 °C
Density: 1.689
Flash Pt: 335 °C
MDL Number: MFCD16619131
CAS Number: 827-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR