2-(2-Bromo-6-nitrophenyl)ethanol ≥97%

Supplier: Matrix Scientific

071166-5G
102519-552EA 946.15 USD
102519-552
2-(2-Bromo-6-nitrophenyl)ethanol ≥97%
2-(2-Bromo-6-nitrophenyl)ethanol

Hazard: Irritant

Formula: C₈H₈BrNO₃
MDL Number: MFCD09834352
CAS Number: 118665-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR