6-Nitro-3-oxo-3,4-dihydropyrazine-2-carboxamide ≥97%

Supplier: Matrix Scientific

071158-5G
102519-544EA 946.15 USD
102519-544
6-Nitro-3-oxo-3,4-dihydropyrazine-2-carboxamide ≥97%
6-Nitro-3-oxo-3,4-dihydropyrazine-2-carboxamide

Hazard: Irritant

Formula: C₅H₄N₄O₄
MDL Number: MFCD21648235
CAS Number: 259793-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR