Methyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-Nitrouracil-6-carboxylate

071126-5G
102519-510EA 946.15 USD
102519-510
Methyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate ≥97%
Methyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₆H₅N₃O₆
MW: 215.12 g/mol
Density: 1.68
MDL Number: MFCD01346736
CAS Number: 6311-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR