4,6-Dichloro-N-methylpyrimidin-2-amine ≥97%

Supplier: Matrix Scientific

071120-5G
102519-502EA 926.15 USD
102519-502
4,6-Dichloro-N-methylpyrimidin-2-amine ≥97%
4,6-Dichloro-N-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₅Cl₂N₃
Boiling Pt: 296 °C
Density: 1.492
Flash Pt: 132 °C
MDL Number: MFCD02091109
CAS Number: 10397-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR