Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-amino-2-(methylthio)pyrimidin-5-carboxylate

071114-5G
102519-494EA 949.23 USD
102519-494
Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate ≥97%
Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₈H₁₁N₃O₂S
MW: 213.2568 g/mol
MDL Number: MFCD00090781
CAS Number: 776-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR