4,4'-(6-Chloropyrimidine-2,4-diyl)dimorpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[4-Chloro-6-(morpholin-4-yl)pyrimidin-2-yl]morpholine

071111-1G
102519-490EA 304.62 USD
102519-490
4,4'-(6-Chloropyrimidine-2,4-diyl)dimorpholine ≥95%
4,4'-(6-Chloropyrimidine-2,4-diyl)dimorpholine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₄O₂
MW: 284.75 g/mol
MDL Number: MFCD01873150
CAS Number: 10244-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR