Methyl-3-amino-5-bromothiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-5-bromothiophene-2-carboxylate

071101-5G
102519-468EA 1269.23 USD
102519-468
Methyl-3-amino-5-bromothiophene-2-carboxylate ≥97%
Methyl-3-amino-5-bromothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₆BrNO₂S
MW: 236.09 g/mol
MDL Number: MFCD12031265
CAS Number: 107818-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR