1-(2-Chloropyridin-3-yl)ethanol ≥97%

Supplier: Matrix Scientific

071094-5G
102519-452EA 1238.46 USD
102519-452
1-(2-Chloropyridin-3-yl)ethanol ≥97%
1-(2-Chloropyridin-3-yl)ethanol

Hazard: Irritant

Formula: C₇H₈ClNO
MDL Number: MFCD16037360
CAS Number: 131674-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR