2,6-Dibromo-4-iodopyridin-3-ol ≥97%

Supplier: Matrix Scientific

071092-5G
102519-448EA 950.77 USD
102519-448
2,6-Dibromo-4-iodopyridin-3-ol ≥97%
2,6-Dibromo-4-iodopyridin-3-ol

Hazard: Irritant

Formula: C₅H₂Br₂INO
MDL Number: MFCD21648228
CAS Number: 478148-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR