2-Bromo-5-methylpyridin-4-amine ≥97%

Supplier: Matrix Scientific

071089-5G
102519-444EA 1980 USD
102519-444
2-Bromo-5-methylpyridin-4-amine ≥97%
2-Bromo-5-methylpyridin-4-amine

Hazard: Irritant

Formula: C₆H₇BrN₂
MDL Number: MFCD11100653
CAS Number: 79055-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR