tert-Butyl-2-methoxypyridin-3-ylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (2-methoxypyridin-3-yl)carbamate

071076-5G
102519-434EA 949.23 USD
102519-434
tert-Butyl-2-methoxypyridin-3-ylcarbamate ≥97%
tert-Butyl-2-methoxypyridin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
MDL Number: MFCD11100821
CAS Number: 161117-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR