Morpholine, 4-[(2R)-oxiranylmethyl](9CI) ≥97%

Supplier: Matrix Scientific

071303-1G
102519-704EA 307.69 USD
102519-704
Morpholine, 4-[(2R)-oxiranylmethyl](9CI) ≥97%
Morpholine, 4-[(2R)-oxiranylmethyl](9CI)

Hazard: Irritant

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD18830136
CAS Number: 452105-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR