Methyl-2-oxotetrahydrofuran-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-oxotetrahydrofuran-3-carboxylate

071292-5G
102519-686EA 692.31 USD
102519-686
Methyl-2-oxotetrahydrofuran-3-carboxylate ≥97%
Methyl-2-oxotetrahydrofuran-3-carboxylate

Hazard: Irritant

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD10569624
CAS Number: 19406-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR