(2S,3S)-3-Hydroxypyrrolidine-2-carboxamide ≥97%

Supplier: Matrix Scientific

071287-5G
102519-678EA 946.15 USD
102519-678
(2S,3S)-3-Hydroxypyrrolidine-2-carboxamide ≥97%
(2S,3S)-3-Hydroxypyrrolidine-2-carboxamide

Hazard: Irritant

Formula: C₅H₁₀N₂O₂
MDL Number: MFCD18831604
CAS Number: 412279-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR