N-(2-Oxoheptan-3-yl)acetamide ≥97%

Supplier: Matrix Scientific

071280-5G
102519-672EA 946.15 USD
102519-672
N-(2-Oxoheptan-3-yl)acetamide ≥97%
N-(2-Oxoheptan-3-yl)acetamide

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD19685805
CAS Number: 30057-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR