Ethyl-3-chloro-2,4-dioxopentanoate ≥97%

Supplier: Matrix Scientific

071278-5G
102519-668EA 946.15 USD
102519-668
Ethyl-3-chloro-2,4-dioxopentanoate ≥97%
Ethyl-3-chloro-2,4-dioxopentanoate

Hazard: Irritant

Formula: C₇H₉ClO₄
MW: 192.6 g/mol
MDL Number: MFCD00173820
CAS Number: 34959-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR