2-Chloro-N-methoxy-N-methylacetamide ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate

071276-5G
102519-664EA 115.38 USD
102519-664
2-Chloro-N-methoxy-N-methylacetamide ≥97%
2-Chloro-N-methoxy-N-methylacetamide

Hazard: Irritant

Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
Boiling Pt: 94…95 °C (9 mmHg)
Melting Pt: 39…41 °C
Flash Pt: 104 °C
MDL Number: MFCD00134232
CAS Number: 67442-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR