2,3-Dimethyl-2-(trimethylsilyloxy)butanenitrile ≥97%

Supplier: Matrix Scientific

071274-5G
102519-660EA 946.15 USD
102519-660
2,3-Dimethyl-2-(trimethylsilyloxy)butanenitrile ≥97%
2,3-Dimethyl-2-(trimethylsilyloxy)butanenitrile

Hazard: Irritant

Formula: C₉H₁₉NOSi
MDL Number: MFCD16786524
CAS Number: 883726-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR