6-Bromo-3-iodoimidazo[1,2-a]pyridine

071063-250MG
102519-414EA 333.73 USD
102519-414
6-Bromo-3-iodoimidazo[1,2-a]pyridine
6-Bromo-3-iodoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄BrIN₂
MW: 322.9300 g/mol
MDL Number: MFCD18800553
CAS Number: 474706-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR