(1H-Pyrrolo[2,3-b]pyridin-6-yl)methanol

Supplier: Matrix Scientific

071041-250MG
102519-370EA 860 USD
102519-370
(1H-Pyrrolo[2,3-b]pyridin-6-yl)methanol
(1H-Pyrrolo[2,3-b]pyridin-6-yl)methanol

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD20486733
CAS Number: 1263413-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR