5-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine
Supplier: Matrix Scientific
071014-250MG
102519-316EA
709.23
USD
102519-316
5-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine
5-Chloro-3-iodo-1H-pyrrolo[3,2-b]pyridine
Hazard: Irritant
Formula:
C₇H₄ClIN₂ |
MDL Number:
MFCD12962967 CAS Number: 1190310-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...