3,5-Dibromo-4-chloropyridine

Supplier: Matrix Scientific

070984-250MG
102519-256EA 709.23 USD
102519-256
3,5-Dibromo-4-chloropyridine
3,5-Dibromo-4-chloropyridine

Hazard: Irritant

Formula: C₅H₂Br₂ClN
MDL Number: MFCD00233993
CAS Number: 13626-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR