5-Bromo-6-chloro-3-iodopyridin-2-amine

Supplier: Matrix Scientific

070980-250MG
102519-248EA 492.31 USD
102519-248
5-Bromo-6-chloro-3-iodopyridin-2-amine
5-Bromo-6-chloro-3-iodopyridin-2-amine

Hazard: Irritant

Formula: C₅H₃BrClIN₂
MDL Number: MFCD20486994
CAS Number: 1207625-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR