6-Chloro-2-iodo-3-methylpyridine

Supplier: Matrix Scientific

070978-250MG
102519-244EA 709.23 USD
102519-244
6-Chloro-2-iodo-3-methylpyridine
6-Chloro-2-iodo-3-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClIN
MDL Number: MFCD18415733
CAS Number: 1261473-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR