6-Chloro-5-iodopyridin-2-amine

Supplier: Matrix Scientific

070976-250MG
102519-240EA 215.38 USD
102519-240
6-Chloro-5-iodopyridin-2-amine
6-Chloro-5-iodopyridin-2-amine

Hazard: Irritant

Formula: C₅H₄ClIN₂
MDL Number: MFCD20486991
CAS Number: 1221398-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR