2-Bromo-4,5-dichloro-6-iodo-3-methoxypyridine

Supplier: Matrix Scientific

070973-250MG
102519-234EA 860 USD
102519-234
2-Bromo-4,5-dichloro-6-iodo-3-methoxypyridine
2-Bromo-4,5-dichloro-6-iodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₃BrCl₂INO
MDL Number: MFCD20487070
CAS Number: 1305324-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR