Ethyl tetrahydropyran-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl tetrahydro-2H-pyran-4-carboxylate

071296-1G
102519-690EA 53.85 USD
102519-690
Ethyl tetrahydropyran-4-carboxylate ≥97%
Ethyl tetrahydropyran-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD08702763
CAS Number: 96835-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR