(2R,3R)-Diethyl-2-acetoxy-3-bromosuccinate ≥97%

Supplier: Matrix Scientific

071281-1G
102519-674EA 250.77 USD
102519-674
(2R,3R)-Diethyl-2-acetoxy-3-bromosuccinate ≥97%
(2R,3R)-Diethyl-2-acetoxy-3-bromosuccinate

Hazard: Irritant

Formula: C₁₀H₁₅BrO₆
MDL Number: MFCD21648266
CAS Number: 74213-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR