2,3-Pyrazinedicarboxylic anhydride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3-Pyrazinecarboxylic anhydride

070860-5G
102519-004EA 27.69 USD
102519-004
2,3-Pyrazinedicarboxylic anhydride ≥98%
2,3-Pyrazinedicarboxylic anhydride

Hazard: Irritant

Formula: C₆H₂N₂O₃
MW: 150.09 g/mol
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00179418
CAS Number: 4744-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR