α-Bromo-2-chlorophenylacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-2-(2-chlorophenyl)acetic acid

070858-25G
102518-998EA 36.92 USD
102518-998
α-Bromo-2-chlorophenylacetic acid ≥98%
α-Bromo-2-chlorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD03428513
CAS Number: 29270-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR