3-Amino-2-bromo-4-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-bromo-4-picoline

070854-1G 070854-5G
102518-978EA 33.85 USD
102518-978 102518-980
3-Amino-2-bromo-4-methylpyridine ≥98%
3-Amino-2-bromo-4-methylpyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD04112497
CAS Number: 126325-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR