1-Chloro-2,5-bis(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
070845-1G
102518-958EA
89.23
USD
102518-958
1-Chloro-2,5-bis(trifluoromethyl)benzene ≥95%
1-Chloro-2,5-bis(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₈H₃ClF₆ |
MDL Number:
MFCD00632388 CAS Number: 328-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...