Methyl-6-(hydroxymethyl)picolinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 6-(hydroxymethyl)picolinateMethyl 6-(hydroxymethyl)pyridine-2-carboxylate

070838-5G
102518-950EA 803.85 USD
102518-950
Methyl-6-(hydroxymethyl)picolinate ≥97%
Methyl 6-(hydroxymethyl)picolinate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 327 °C
Density: 1.244
Flash Pt: 151 °C
MDL Number: MFCD03840850
CAS Number: 39977-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR