4-(3-(Trifluoromethyl)phenyl)pyridine ≥98%

Supplier: Matrix Scientific

070819-1G
102518-930EA 164.62 USD
102518-930
4-(3-(Trifluoromethyl)phenyl)pyridine ≥98%
4-(3-(Trifluoromethyl)phenyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈F₃N
MDL Number: MFCD15475192
CAS Number: 5957-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR