3-Pyrrolidinecarboxamide ≥95%

Supplier: Matrix Scientific

069191-1G
102833-456EA 689.23 USD
102833-456
3-Pyrrolidinecarboxamide ≥95%
3-Pyrrolidinecarboxamide

Hazard: Irritant

Formula: C₅H₁₀N₂O
MDL Number: MFCD09037932
CAS Number: 471254-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR