tert-Butyl-4-formylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-formylpiperidineN-Boc-4-Formylpiperidine1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehydeTert-butyl 4-formylpiperidine-1-carboxylate1-(tert-Butoxycarbonyl)-4-formylpiperidinetert-Butyl 4-formylpiperidine-1-carboxylate1-BOC-4-Piperidinecarboxaldehyde1-Boc-4-piperidinecarboxaldehyde

068629-25G
102551-716EA 75.38 USD
102551-716
tert-Butyl-4-formylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-formylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR