Oxetane-3-carboxylic acid

Supplier: Matrix Scientific

066986-1G
102518-906EA 515.38 USD
102518-906
Oxetane-3-carboxylic acid
Oxetane-3-carboxylic acid

Hazard: Irritant/Store in Freezer

Formula: C₄H₆O₃
Boiling Pt: 244.2 °C
Density: 1.374
Flash Pt: 115 °C
MDL Number: MFCD08669505
CAS Number: 114012-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR