3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid

Supplier: Matrix Scientific

062790-1G
102833-320EA 676.92 USD
102833-320
3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid
3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₀Cl₂O₂
Boiling Pt: 290 °C
Density: 1.433
Flash Pt: 129 °C
MDL Number: MFCD00137937
CAS Number: 55701-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR