(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)acetyl chloride

065425-25G
102833-394EA 1315.17 USD
102833-394
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl chloride

Hazard: Irritant

Formula: C₁₀H₆ClNO₃
MW: 223.62 g/mol
MDL Number: MFCD00192872
CAS Number: 6780-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR