2-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride

Supplier: Matrix Scientific

060804-500MG
102663-786EA 752.31 USD
102663-786
2-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride
2-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride

Hazard: Irritant

Formula: C₅H₇Cl₂NS
MDL Number: MFCD18071283
CAS Number: 921145-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR