1-Cyclopropylbutane-1,3-dione

Supplier: Matrix Scientific

065612-5G 065612-25G
102833-408EA 56.92 USD
102833-408 102833-410
1-Cyclopropylbutane-1,3-dione
1-Cyclopropylbutane-1,3-dione

Hazard: Irritant

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 68…70 °C (12 Torr)
MDL Number: MFCD00511094
CAS Number: 21573-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR