Daidzein

Supplier: Matrix Scientific

Synonyms: 4',7-DihydroxyisoflavoneDaidzeol7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone

065243-1G
102551-676EA 235.38 USD
102551-676
Daidzein
Daidzein

Hazard: Irritant

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
MDL Number: MFCD00016954
CAS Number: 486-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR