Methyl-2-pyrazinecarboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl pyrazine-2-carboxylate
, Methyl 2-pyrazinecarboxylate
064371-25G
064371-5G
102551-646EA
118.46
USD
102551-646
102551-648
Methyl-2-pyrazinecarboxylate
Methyl 2-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00014611 CAS Number: 6164-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...