4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
Supplier: Matrix Scientific
Synonyms:
4-Methoxy-1H-pyrrol-2(5H)-one
, 4-Methoxy-3-pyrrolin-2-one
064111-1G
064111-5G
102833-354EA
300
USD
102833-354
102833-352
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
Hazard: Irritant
Formula:
C₅H₇NO₂ MW: 113.12 g/mol |
MDL Number:
MFCD00071564 CAS Number: 69778-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...