3-Chloroisoxazole-5-carboxylic acid

Supplier: Matrix Scientific

063909-500MG 063909-2.5G
102518-898EA 573.85 USD
102518-898 102518-900
3-Chloroisoxazole-5-carboxylic acid
3-Chloroisoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₄H₂ClNO₃
MDL Number: MFCD01716783
CAS Number: 20724-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR