2-Formyl-5-picoline
Supplier: Matrix Scientific
Synonyms:
5-Methylpicolinaldehyde
, 5-Methylpyridine-2-carbaldehyde
063862-5G
102551-642EA
113.85
USD
102551-642
2-Formyl-5-picoline
2-Formyl-5-picoline
Hazard: Irritant
Formula:
C₇H₇NO MW: 121.1400 g/mol |
MDL Number:
MFCD07367942 CAS Number: 4985-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...