Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide

Supplier: Matrix Scientific

Synonyms: 4-Hydroxytetrahydro-2H-thiopyran 1,1-dioxide

062650-1G
102518-892EA 369.23 USD
102518-892
Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide
Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide

Hazard: Irritant

Formula: C₅H₁₀O₃S
MW: 150.2 g/mol
MDL Number: MFCD12152579
CAS Number: 194152-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR