4-Methyl-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

061226-1G
102551-576EA 584.62 USD
102551-576
4-Methyl-1,2,5-oxadiazol-3-amine
4-Methyl-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₃H₅N₃O
MDL Number: MFCD00186403
CAS Number: 17647-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR